missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethyl Methyl Ether 98.0+%, TCI America™
$70.84 - $346.71
Chemical Identifiers
| CAS | 3558-60-9 |
|---|---|
| Molecular Formula | C9H12O |
| Molecular Weight (g/mol) | 136.194 |
| MDL Number | MFCD00025912 |
| InChI Key | CQLYXIUHVFRXLT-UHFFFAOYSA-N |
| Synonym | Methyl Phenethyl Ether |
| PubChem CID | 19089 |
| IUPAC Name | 2-methoxyethylbenzene |
| SMILES | COCCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P126325ML
|
TCI America
P126325ML |
25 mL |
Each for $70.84
|
|
|||||
|
P1263500ML
|
TCI America
P1263500ML |
500 mL |
Each for $346.71
|
|
|||||
Chemical Identifiers
| 3558-60-9 | |
| 136.194 | |
| CQLYXIUHVFRXLT-UHFFFAOYSA-N | |
| 19089 | |
| COCCC1=CC=CC=C1 |
| C9H12O | |
| MFCD00025912 | |
| Methyl Phenethyl Ether | |
| 2-methoxyethylbenzene |
Specifications
| 3558-60-9 | |
| 190°C | |
| MFCD00025912 | |
| Methyl Phenethyl Ether | |
| COCCC1=CC=CC=C1 | |
| 136.194 | |
| 136.19 | |
| Liquid |
| Yellow | |
| C9H12O | |
| 25 mL | |
| CQLYXIUHVFRXLT-UHFFFAOYSA-N | |
| 2-methoxyethylbenzene | |
| 19089 | |
| ≥98.0% (GC) | |
| 2-Phenylethyl Methyl Ether |
Safety and Handling
EINECSNumber : (3)-1035
RTECSNumber : KO2230000
TSCA : Yes