missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Cinnamate 99.0+%, TCI America™

$44.73 - $164.03

Chemical Identifiers

CAS 103-53-7
Molecular Formula C17H16O2
Molecular Weight (g/mol) 252.313
MDL Number MFCD00022050
InChI Key MJQVZIANGRDJBT-VAWYXSNFSA-N
Synonym Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester
PubChem CID 5369459
IUPAC Name 2-phenylethyl (E)-3-phenylprop-2-enoate
SMILES C1=CC=C(C=C1)CCOC(=O)C=CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P200725G
View Documents
TCI America
P200725G
25 g N/A
Only null left
 
P2007500G
View Documents
TCI America
P2007500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

103-53-7
252.313
MJQVZIANGRDJBT-VAWYXSNFSA-N
5369459
C1=CC=C(C=C1)CCOC(=O)C=CC2=CC=CC=C2
C17H16O2
MFCD00022050
Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester
2-phenylethyl (E)-3-phenylprop-2-enoate

Specifications

103-53-7
White-Yellow
MFCD00022050
Cinnamic Acid 2-Phenylethyl Ester, Phenethyl Cinnamate, Cinnamic Acid Phenethyl Ester
C1=CC=C(C=C1)CCOC(=O)C=CC2=CC=CC=C2
252.313
252.31
Crystalline Powder
57°C
C17H16O2
25 g
MJQVZIANGRDJBT-VAWYXSNFSA-N
2-phenylethyl (E)-3-phenylprop-2-enoate
5369459
≥99.0% (GC)
2-Phenylethyl Cinnamate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1739

RTECSNumber : GE0405000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'