missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl Acetate 98.0+%, TCI America™

Catalog No. p-7117008
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A069225G 25 g
A0692500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A069225G Supplier TCI America Supplier No. A069225G
Only null left

Chemical Identifiers

CAS 103-45-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008720
InChI Key MDHYEMXUFSJLGV-UHFFFAOYSA-N
Synonym phenethyl acetate, 2-phenethyl acetate, acetic acid, 2-phenylethyl ester, benzylcarbinyl acetate, beta-phenylethyl acetate, acetic acid, phenethyl ester, phenethyl alcohol, acetate, phenylethyl acetate, acetic acid phenethyl ester, ethanol, 2-phenyl-, acetate
PubChem CID 7654
ChEBI CHEBI:31988
IUPAC Name 2-phenylethyl acetate
SMILES CC(=O)OCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 224°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenylethyl Acetate
EINECSNumber (3)-1040&(3)-1045
RTECSNumber AJ2220000
TSCA Yes