missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl-1-boronic acid pinacol ester, 99%

Catalog No. aal19530md
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19530MD 250 mg
AAL1953003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19530MD Supplier Thermo Scientific Chemicals Supplier No. L19530MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 165904-22-3 | C14H21BO2 | 232.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165904-22-3
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD03788721
InChI Key LVLQNRWCBBIVHR-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-phenethyl-1,3,2-dioxaborolane, 2-phenylethyl-1-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylethyl-1,3,2-dioxaborolane, 2-phenylethylboronic acid, pinacol ester, phenethylboronic acid pinacol ester, 2-phenylethylboronic acid pinacol ester, 2-phenylethylboronic acid,pinacol ester, 2-phenylethyl boronic acid pinacol ester, 2-phenyl ethyl-1-boronic acid pinacol ester, 2-phenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11333720
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylethyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC=CC=C2

Specifications

Melting Point 31°C to 33°C
Boiling Point 70°C to 80°C
Refractive Index 1.486
Quantity 250 mg
Beilstein 8258927
Formula Weight 232.13
Percent Purity 99%
Chemical Name or Material 2-Phenylethyl-1-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only