missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanethiol 97.0+%, TCI America™

Catalog No. P17155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17155G 5 g
P171525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17155G Supplier TCI America Supplier No. P17155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4410-99-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00004891
InChI Key ZMRFRBHYXOQLDK-UHFFFAOYSA-N
Synonym phenethyl mercaptan, benzeneethanethiol, 2-phenylethyl mercaptan, phenylethyl mercaptan, 2-phenylethylthiol, unii-i4th2t2iny, 2-phenyl-ethanethiol, 2-phenylethylmercaptan, beta-phenylethyl mercaptan, i4th2t2iny
PubChem CID 78126
IUPAC Name 2-phenylethane-1-thiol
SMILES SCCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 218°C
Quantity 5 g
Formula Weight 138.23
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Phenylethanethiol