missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanethiol, 99%

Catalog No. p-3779427
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378490100 10 g Glass bottle
AC378490500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378490100 Supplier Thermo Scientific Chemicals Supplier No. 378490100

CAS: 4410-99-5 | C8H10S | 138.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4410-99-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.23
MDL Number MFCD00004891
InChI Key ZMRFRBHYXOQLDK-UHFFFAOYSA-N
Synonym phenethyl mercaptan, benzeneethanethiol, 2-phenylethyl mercaptan, phenylethyl mercaptan, 2-phenylethylthiol, unii-i4th2t2iny, 2-phenyl-ethanethiol, 2-phenylethylmercaptan, beta-phenylethyl mercaptan, i4th2t2iny
PubChem CID 78126
SMILES SCCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0300g/mL
Boiling Point 97°C to 100°C (12.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.561
Linear Formula C6H5(CH2)2SH
Quantity 10 g
Specific Gravity 1.03
IUPAC Name 2-phenylethane-1-thiol
Formula Weight 138.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Phenylethanethiol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-563-9

RUO – Research Use Only