missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylcyclohexanone 98.0+%, TCI America™

Catalog No. P13115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13115G 5 g
P13111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13115G Supplier TCI America Supplier No. P13115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1444-65-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001629
InChI Key DRLVMOAWNVOSPE-UHFFFAOYSA-N
Synonym 2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone
PubChem CID 95592
IUPAC Name 2-phenylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)C2=CC=CC=C2

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylcyclohexanone