missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole 98.0+%, TCI America™

Catalog No. P11055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11055G Supplier TCI America Supplier No. P11055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 716-79-0
Molecular Formula C13H10N2
Quantity 5 g
MDL Number MFCD00005592
Synonym 2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole
InChI Key DWYHDSLIWMUSOO-UHFFFAOYSA-N
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=C1
IUPAC Name 2-phenyl-1H-1,3-benzodiazole
Molecular Weight (g/mol) 194.24
PubChem CID 12855
Formula Weight 194.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylbenzimidazole
Show More Show Less
RTECSNumber DE0360000
TSCA Yes