missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole, 97%

Catalog No. AAA1099622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1099622 100 g
AAA1099606 5 g
AAA1099614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1099622 Supplier Thermo Scientific Chemicals Supplier No. A1099622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 716-79-0 | C13H10N2 | 194.24 g/mol

2-Phenylbenzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylbenzimidazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 716-79-0
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
MDL Number MFCD00005592
InChI Key DWYHDSLIWMUSOO-UHFFFAOYSA-N
Synonym 2-phenylbenzimidazole, 2-phenyl-1h-benzo d imidazole, phenizidole, phenzidole, phenzidol, gainex, 1h-benzimidazole, 2-phenyl, benzimidazole, 2-phenyl, 2-phenylbenzimdazole, 2-phenyl-1h-benzoimidazole
PubChem CID 12855
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC=C1

Specifications

Melting Point 296°C to 297°C
Quantity 100 g
Beilstein 7087
Merck Index 14,7274
Solubility Information Slightly soluble in water.
IUPAC Name 2-phenyl-1H-benzimidazole
Formula Weight 194.24
Percent Purity 97%
Chemical Name or Material 2-Phenylbenzimidazole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-939-2
RTECSNumber DE0360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only