missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole-5-sulfonic acid, 98%

Catalog No. AC259702500
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259702500 250 g Glass bottle
AC259700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC259702500 Supplier Thermo Scientific Chemicals Supplier No. 259702500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27503-81-7 | C13H10N2O3S | 274.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27503-81-7
Molecular Formula C13H10N2O3S
Molecular Weight (g/mol) 274.29
MDL Number MFCD00053007
InChI Key UVCJGUGAGLDPAA-UHFFFAOYSA-N
Synonym ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid
PubChem CID 33919
IUPAC Name 2-phenyl-3H-benzimidazole-5-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 g
Merck Index 15, 3647
Solubility Information Solubility in water: soluble
Formula Weight 274.28
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Phenylbenzimidazole-5-sulfonic acid
Show More Show Less
EINECSNumber 248-502-

RUO – Research Use Only