missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylbenzimidazole-5-sulfonic acid, 98%
CAS: 27503-81-7 | C13H10N2O3S | 274.29 g/mol
$80.13 - $80.13
Chemical Identifiers
| CAS | 27503-81-7 |
|---|---|
| Molecular Formula | C13H10N2O3S |
| Molecular Weight (g/mol) | 274.29 |
| MDL Number | MFCD00053007 |
| InChI Key | UVCJGUGAGLDPAA-UHFFFAOYSA-N |
| Synonym | ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid |
| PubChem CID | 33919 |
| IUPAC Name | 2-phenyl-3H-benzimidazole-5-sulfonic acid |
| SMILES | OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC259700250
|
Thermo Scientific Chemicals
259700250 |
25 g | Glass bottle |
Each for $80.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 27503-81-7 | |
| 274.29 | |
| UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
| 33919 | |
| OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 |
| C13H10N2O3S | |
| MFCD00053007 | |
| ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid | |
| 2-phenyl-3H-benzimidazole-5-sulfonic acid |
Specifications
| 27503-81-7 | |
| 100.0 | |
| White | |
| 98% | |
| C13H10N2O3S | |
| 25 g | |
| ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid | |
| UVCJGUGAGLDPAA-UHFFFAOYSA-N | |
| 2-phenyl-3H-benzimidazole-5-sulfonic acid | |
| 33919 | |
| 98% | |
| 2-Phenylbenzimidazole-5-sulfonic acid |
| 97.5 | |
| >300°C | |
| Authentic | |
| Glass bottle | |
| MFCD00053007 | |
| 15, 3647 | |
| Solubility in water: soluble | |
| OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1 | |
| 274.29 | |
| 274.28 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 248-502-
RUO – Research Use Only