missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-9H-carbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 290°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 243.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Phenyl-9H-carbazole |
Safety and Handling
| TSCA | No |