missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-9H-carbazole 98.0+%, TCI America™

Catalog No. P23751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23751G 1 g
P23755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23751G Supplier TCI America Supplier No. P23751G
Only null left

Chemical Identifiers

CAS 88590-00-5
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.309
MDL Number MFCD26402145
InChI Key IMLDYQBWZHPGJA-UHFFFAOYSA-N
PubChem CID 49799641
IUPAC Name 2-phenyl-9H-carbazole
SMILES C1=CC=C(C=C1)C2=CC3=C(C=C2)C4=CC=CC=C4N3

Specifications

Melting Point 290°C
Color White-Yellow
Quantity 1 g
Formula Weight 243.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenyl-9H-carbazole