Learn More
2-Phenyl-2-imidazoline, 98+%, Thermo Scientific™
$102.44 - $102.44
Chemical Identifiers
| CAS | 936-49-2 |
|---|---|
| Molecular Formula | C9H10N2 |
| Molecular Weight (g/mol) | 146.19 |
| MDL Number | MFCD00005180 |
| InChI Key | BKCCAYLNRIRKDJ-UHFFFAOYSA-N |
| Synonym | 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference |
| PubChem CID | 13639 |
| IUPAC Name | 2-phenyl-4,5-dihydro-1H-imidazole |
| SMILES | C1CN=C(N1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC199731000
|
Thermo Scientific Chemicals
199731000 |
100 g | Glass bottle |
Each for $102.44
|
|
||||
Chemical Identifiers
| 936-49-2 | |
| 146.19 | |
| BKCCAYLNRIRKDJ-UHFFFAOYSA-N | |
| 13639 | |
| C1CN=C(N1)C1=CC=CC=C1 |
| C9H10N2 | |
| MFCD00005180 | |
| 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference | |
| 2-phenyl-4,5-dihydro-1H-imidazole |
Specifications
| 936-49-2 | |
| 100.0 | |
| Yellow | |
| 201°C | |
| 98+% | |
| C9H10N2 | |
| 100 g | |
| 2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference | |
| BKCCAYLNRIRKDJ-UHFFFAOYSA-N | |
| 2-phenyl-4,5-dihydro-1H-imidazole | |
| 13639 | |
| 98+% | |
| 2-Phenyl-2-imidazoline, 98+% |
| 98.0 | |
| 94°C to 99°C | |
| 188°C (20.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| MFCD00005180 | |
| 23, V,6, 425 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohols | |
| C1CN=C(N1)C1=CC=CC=C1 | |
| 146.19 | |
| 146.19 | |
| Crystalline Powder or Flakes |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 213-313-4
RTECSNumber : NJ4395500
TSCA : TSCA
RUO – Research Use Only