missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-1-propanol 98.0+%, TCI America™
$78.29 - $512.62
Chemical Identifiers
| CAS | 1123-85-9 |
|---|---|
| Molecular Formula | C9H12O |
| Molecular Weight (g/mol) | 136.194 |
| MDL Number | MFCD00004736 |
| InChI Key | RNDNSYIPLPAXAZ-UHFFFAOYSA-N |
| Synonym | 2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl |
| PubChem CID | 14295 |
| IUPAC Name | 2-phenylpropan-1-ol |
| SMILES | CC(CO)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P081325ML
|
TCI America
P081325ML |
25 mL |
Each for $78.29
|
|
|||||
|
P0813500ML
|
TCI America
P0813500ML |
500 mL |
Each for $512.62
|
|
|||||
Chemical Identifiers
| 1123-85-9 | |
| 136.194 | |
| RNDNSYIPLPAXAZ-UHFFFAOYSA-N | |
| 14295 | |
| CC(CO)C1=CC=CC=C1 |
| C9H12O | |
| MFCD00004736 | |
| 2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl | |
| 2-phenylpropan-1-ol |
Specifications
| 1123-85-9 | |
| 223°C | |
| MFCD00004736 | |
| 2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl | |
| CC(CO)C1=CC=CC=C1 | |
| 136.194 | |
| 136.19 | |
| Liquid |
| Colorless | |
| C9H12O | |
| 25 mL | |
| RNDNSYIPLPAXAZ-UHFFFAOYSA-N | |
| 2-phenylpropan-1-ol | |
| 14295 | |
| ≥98.0% (GC) | |
| 2-Phenyl-1-propanol |
Safety and Handling
EINECSNumber : (3)-1034
RTECSNumber : SG8590000
TSCA : Yes