missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1-propanol 98.0+%, TCI America™

Catalog No. P0813500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0813500ML 500 mL
P081325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0813500ML Supplier TCI America Supplier No. P0813500ML

Chemical Identifiers

CAS 1123-85-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004736
InChI Key RNDNSYIPLPAXAZ-UHFFFAOYSA-N
Synonym 2-phenyl-1-propanol, hydratropic alcohol, hydratropyl alcohol, 2-phenylpropanol-1, beta-methylphenethyl alcohol, 2-phenylpropyl alcohol, 1-propanol, 2-phenyl, 1-hydroxy-2-phenylpropane, 2-phenylpropanol, benzeneethanol, .beta.-methyl
PubChem CID 14295
IUPAC Name 2-phenylpropan-1-ol
SMILES CC(CO)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 223°C
Quantity 500 mL
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenyl-1-propanol
EINECSNumber (3)-1034
RTECSNumber SG8590000
TSCA Yes