missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC18301DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18301DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC18301DA Supplier Thermo Scientific Chemicals Supplier No. CC18301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7113-10-2
Molecular Formula C10H6NO2S
Molecular Weight (g/mol) 204.22
MDL Number MFCD00141954
InChI Key IBUSLNJQKLZPNR-UHFFFAOYSA-M
Synonym 2-phenylthiazole-4-carboxylic acid, 2-phenyl-4-carboxythiazole, 4-thiazolecarboxylic acid, 2-phenyl, 2-phenyl-thiazole-4-carboxylic acid, 2-phenyl-4-thiazolecarboxylic acid, 2-phenyl-1,3-thiadiazole-4-carboxylic acid, pubchem19074, 4-carboxy-2-phenylthiazole, ksc237i8r, phenyl-1,3-thiazole-4-carboxylic acid
PubChem CID 138928

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.