missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenyl-1,3-indandione 98.0+%, TCI America™
$130.48 - $130.48
Chemical Identifiers
| CAS | 83-12-5 |
|---|---|
| Molecular Formula | C15H10O2 |
| Molecular Weight (g/mol) | 222.243 |
| MDL Number | MFCD00003782 |
| InChI Key | NFBAXHOPROOJAW-UHFFFAOYSA-N |
| Synonym | phenindione, 2-phenyl-1,3-indandione, dindevan, 2-phenyl-1h-indene-1,3 2h-dione, phenylindanedione, athrombon, diophindane, emandione, fenhydren, phenhydren |
| PubChem CID | 4760 |
| ChEBI | CHEBI:8066 |
| IUPAC Name | 2-phenylindene-1,3-dione |
| SMILES | C1=CC=C(C=C1)C2C(=O)C3=CC=CC=C3C2=O |
Chemical Identifiers
| 83-12-5 | |
| 222.243 | |
| NFBAXHOPROOJAW-UHFFFAOYSA-N | |
| 4760 | |
| 2-phenylindene-1,3-dione |
| C15H10O2 | |
| MFCD00003782 | |
| phenindione, 2-phenyl-1,3-indandione, dindevan, 2-phenyl-1h-indene-1,3 2h-dione, phenylindanedione, athrombon, diophindane, emandione, fenhydren, phenhydren | |
| CHEBI:8066 | |
| C1=CC=C(C=C1)C2C(=O)C3=CC=CC=C3C2=O |
Specifications
| 83-12-5 | |
| White-Yellow | |
| MFCD00003782 | |
| 2811 | |
| NFBAXHOPROOJAW-UHFFFAOYSA-N | |
| 2-phenylindene-1,3-dione | |
| 4760 | |
| 222.24 | |
| Crystalline Powder |
| 148°C | |
| C15H10O2 | |
| 25 g | |
| phenindione, 2-phenyl-1,3-indandione, dindevan, 2-phenyl-1h-indene-1,3 2h-dione, phenylindanedione, athrombon, diophindane, emandione, fenhydren, phenhydren | |
| C1=CC=C(C=C1)C2C(=O)C3=CC=CC=C3C2=O | |
| 222.243 | |
| CHEBI:8066 | |
| ≥98.0% (GC) | |
| 2-Phenyl-1,3-indandione |
Safety and Handling
RTECSNumber : NK6125000
TSCA : No