missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxypropionyl Chloride 98.0+%, TCI America™
$128.62 - $1549.03
Chemical Identifiers
| CAS | 122-35-0 |
|---|---|
| Molecular Formula | C9H9ClO2 |
| Molecular Weight (g/mol) | 184.619 |
| MDL Number | MFCD00018810 |
| InChI Key | BDSSZTXPZHIYHM-UHFFFAOYSA-N |
| Synonym | 2-phenoxypropionyl chloride, propanoyl chloride, 2-phenoxy, 2-phenoxypropionic acid chloride, alpha-phenoxypropionyl chloride, 2-phenoxy-propionyl chloride, propanoyl chloride,2-phenoxy, propionyl chloride, 2-phenoxy, .alpha.-phenoxypropionyl chloride, acmc-1bvfg, 2-phenoxypropionylchloride |
| PubChem CID | 95436 |
| IUPAC Name | 2-phenoxypropanoyl chloride |
| SMILES | CC(C(=O)Cl)OC1=CC=CC=C1 |
Chemical Identifiers
| 122-35-0 | |
| 184.619 | |
| BDSSZTXPZHIYHM-UHFFFAOYSA-N | |
| 95436 | |
| CC(C(=O)Cl)OC1=CC=CC=C1 |
| C9H9ClO2 | |
| MFCD00018810 | |
| 2-phenoxypropionyl chloride, propanoyl chloride, 2-phenoxy, 2-phenoxypropionic acid chloride, alpha-phenoxypropionyl chloride, 2-phenoxy-propionyl chloride, propanoyl chloride,2-phenoxy, propionyl chloride, 2-phenoxy, .alpha.-phenoxypropionyl chloride, acmc-1bvfg, 2-phenoxypropionylchloride | |
| 2-phenoxypropanoyl chloride |
Specifications
| 122-35-0 | |
| 106°C | |
| MFCD00018810 | |
| 3265 | |
| BDSSZTXPZHIYHM-UHFFFAOYSA-N | |
| 2-phenoxypropanoyl chloride | |
| 95436 | |
| ≥98.0% (GC,T) | |
| 2-Phenoxypropionyl Chloride |
| Yellow | |
| C9H9ClO2 | |
| 25 g | |
| 2-phenoxypropionyl chloride, propanoyl chloride, 2-phenoxy, 2-phenoxypropionic acid chloride, alpha-phenoxypropionyl chloride, 2-phenoxy-propionyl chloride, propanoyl chloride,2-phenoxy, propionyl chloride, 2-phenoxy, .alpha.-phenoxypropionyl chloride, acmc-1bvfg, 2-phenoxypropionylchloride | |
| CC(C(=O)Cl)OC1=CC=CC=C1 | |
| 184.619 | |
| 184.62 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2838&(9)-1147&(9)-1258
TSCA : Yes