missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethylamine, 98%

Catalog No. p-7081999
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1222506 5 g
AAA1222514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1222506 Supplier Thermo Scientific Chemicals Supplier No. A1222506
Only null left

CAS: 1758-46-9 | C8H11NO | 137.182 g/mol

Organic intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1758-46-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00052975
InChI Key IMLAIXAZMVDRGA-UHFFFAOYSA-N
Synonym 2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
PubChem CID 15651
IUPAC Name 2-phenoxyethanamine
SMILES C1=CC=C(C=C1)OCCN

Specifications

Density 1.048
Boiling Point 88°C to 90°C (1 mmHg)
Flash Point 107°C (224°F)
Odor Amine-like
Refractive Index 1.537
Linear Formula C6H5O(CH2)2NH2
Quantity 5 g
UN Number UN2735
Beilstein 774671
Sensitivity Air Sensitive
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 2-Phenoxyethylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 217-153-6
RTECSNumber KR9500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only