missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl Methacrylate (stabilized with HQ + MEHQ) 85.0+%, TCI America™

Catalog No. P2468500G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2468500G 500 g
P2468100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2468500G Supplier TCI America Supplier No. P2468500G
Only null left

Chemical Identifiers

CAS 10595-06-9
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00053685
InChI Key CEXQWAAGPPNOQF-UHFFFAOYSA-N
Synonym Methacrylic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Methacrylate
PubChem CID 66357
IUPAC Name 2-phenoxyethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOC1=CC=CC=C1

Specifications

Color Yellow
pH <7
Boiling Point 260°C
Quantity 500 g
Formula Weight 206.24
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethyl Methacrylate (stabilized with HQ + MEHQ)
EINECSNumber (3)-4214
RTECSNumber 3286610
TSCA Yes