missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethyl Acrylate (stabilized with MEHQ) 93.0+%, TCI America™

Catalog No. A1400500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1400500G 500 g
A140025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1400500G Supplier TCI America Supplier No. A1400500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 48145-04-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00053696
InChI Key RZVINYQDSSQUKO-UHFFFAOYSA-N
Synonym 2-phenoxyethyl acrylate, phenoxyethyl acrylate, light ester po-a, chemlink 160, ebecryl 110, 2-propenoic acid, 2-phenoxyethyl ester, ethylene glycol phenyl ether acrylate, 2-phenoxyethanol acrylate, phenyl cellosolve acrylate, acrylic acid, 2-phenoxyethyl ester
PubChem CID 39485
IUPAC Name 2-phenoxyethyl prop-2-enoate
SMILES C=CC(=O)OCCOC1=CC=CC=C1

Specifications

Color Yellow
Quantity 500 g
UN Number 3082
Formula Weight 192.21
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethyl Acrylate (stabilized with MEHQ)
EINECSNumber (3)-3684
RTECSNumber KM0700000
TSCA Yes