missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxyethyl Acrylate (stabilized with MEHQ) 93.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| UN Number | 3082 |
| Formula Weight | 192.21 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Phenoxyethyl Acrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (3)-3684 |
| RTECSNumber | KM0700000 |
| TSCA | Yes |