missing translation for 'onlineSavingsMsg'
Learn More

2-Pentynoic acid, 97%

CAS: 5963-77-9 | C5H6O2 | 98.101 g/mol

$61.10 - $727.73

Chemical Identifiers

CAS 5963-77-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00014370
InChI Key MINRDQDGBLQBGD-UHFFFAOYSA-N
Synonym 2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
PubChem CID 324379
IUPAC Name pent-2-ynoic acid
SMILES CCC#CC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5341103
View Documents
Thermo Scientific Chemicals
H5341103
1 g
Each for $61.10
Only null left
 
AAH5341106
View Documents
Thermo Scientific Chemicals
H5341106
5 g
Each for $210.66
Only null left
 
AAH5341114
View Documents
Thermo Scientific Chemicals
H5341114
25 g
Each for $727.73
Only null left
 
Description

Description

2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5963-77-9
98.101
MINRDQDGBLQBGD-UHFFFAOYSA-N
324379
CCC#CC(=O)O
C5H6O2
MFCD00014370
2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
pent-2-ynoic acid

Specifications

5963-77-9
96°C (205°F)
CH3CH2CH≡CCO2H
1 g
MINRDQDGBLQBGD-UHFFFAOYSA-N
pent-2-ynoic acid
324379
97%
47°C to 53°C
C5H6O2
MFCD00014370
2-pentynoic acid, pentynoic acid, 2-pentynoicacid, 2-pentyneoic acid, acmc-1ay8s
CCC#CC(=O)O
98.101
98.1
2-Pentynoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only