missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyne 97.0+%, TCI America™

Catalog No. p-7134704
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06285ML 5 mL
P062825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06285ML Supplier TCI America Supplier No. P06285ML
Only null left

Chemical Identifiers

CAS 627-21-4
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009380
InChI Key NKTDTMONXHODTI-UHFFFAOYSA-N
Synonym 2-pentyne, ethylmethylacetylene, unii-57ng6hki9d, 1-ethyl-2-methylacetylene, 57ng6hki9d, acmc-209n7k, c2h5c.$.cch3, 2-pentyne 6ci,7ci,8ci,9ci
PubChem CID 12310
IUPAC Name pent-2-yne
SMILES CCC#CC

Specifications

Color Yellow
Boiling Point 56°C
Quantity 5 mL
UN Number 3295
Formula Weight 68.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentyne