missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyne, 98+%

Catalog No. AAL0297314
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0297314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0297314 Supplier Thermo Scientific Chemicals Supplier No. L0297314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-21-4 | C5H8 | 68.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-21-4
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009380
InChI Key NKTDTMONXHODTI-UHFFFAOYSA-N
Synonym 2-pentyne, ethylmethylacetylene, unii-57ng6hki9d, 1-ethyl-2-methylacetylene, 57ng6hki9d, acmc-209n7k, c2h5c.$.cch3, 2-pentyne 6ci,7ci,8ci,9ci
PubChem CID 12310
IUPAC Name pent-2-yne
SMILES CCC#CC

Specifications

Melting Point -109°C
Density 0.71
Boiling Point 55°C to 56°C
Flash Point −20°C (−4°F)
Odor Unpleasant
Refractive Index 1.403
Quantity 25 g
UN Number UN3295
Beilstein 1696938
Formula Weight 68.12
Percent Purity ≥98%
Chemical Name or Material 2-Pentyne
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 210-989-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only