missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentyn-1-ol 98.0+%, TCI America™
Supplier: TCI America P081525ML
Specifications
| 2-Pentyn-1-ol | |
| Yellow | |
| C5H8O | |
| 25 mL | |
| WLPYSOCRPHTIDZ-UHFFFAOYSA-N | |
| pent-2-yn-1-ol | |
| 80421 | |
| ≥98.0% (GC) |
| 6261-22-9 | |
| 154°C | |
| MFCD00040915 | |
| 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01 | |
| CCC#CCO | |
| 84.118 | |
| 84.12 | |
| Liquid |
Chemical Identifiers
| 6261-22-9 | |
| 84.118 | |
| WLPYSOCRPHTIDZ-UHFFFAOYSA-N | |
| 80421 | |
| CCC#CCO |
| C5H8O | |
| MFCD00040915 | |
| 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01 | |
| pent-2-yn-1-ol |
Safety and Handling
EINECSNumber : (2)-3355
TSCA : Yes