missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyn-1-ol 98.0+%, TCI America™

Catalog No. P08155ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08155ML 5 mL
P081525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08155ML Supplier TCI America Supplier No. P08155ML
Only null left

Chemical Identifiers

CAS 6261-22-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00040915
InChI Key WLPYSOCRPHTIDZ-UHFFFAOYSA-N
Synonym 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01
PubChem CID 80421
IUPAC Name pent-2-yn-1-ol
SMILES CCC#CCO

Specifications

Color Yellow
Boiling Point 154°C
Quantity 5 mL
Formula Weight 84.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentyn-1-ol
EINECSNumber (2)-3355
TSCA Yes