missing translation for 'onlineSavingsMsg'
Learn More

2-Pentyn-1-ol, 98%

Catalog No. AAA1299903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299903 1 g
AAA1299906 5 g
AAA1299914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1299903 Supplier Thermo Scientific Chemicals Supplier No. A1299903
Only null left

CAS: 6261-22-9 | C5H8O | 84.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6261-22-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00040915
InChI Key WLPYSOCRPHTIDZ-UHFFFAOYSA-N
Synonym 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01
PubChem CID 80421
IUPAC Name pent-2-yn-1-ol
SMILES CCC#CCO

Specifications

Density 0.91
Boiling Point 154°C
Flash Point 58°C (136°F)
Odor Recognizable
Refractive Index 1.453
Linear Formula CH3CH2C≡CCH2OH
Quantity 1 g
UN Number UN1987
Beilstein 1734007
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 2-Pentyn-1-ol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 228-411-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only