missing translation for 'onlineSavingsMsg'
Learn More

2-Pentylthiophene 98.0+%, TCI America™

Catalog No. P18355G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18355G Supplier TCI America Supplier No. P18355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4861-58-9
Molecular Formula C9H14S
Molecular Weight (g/mol) 154.271
MDL Number MFCD00041017
InChI Key NOYVOSGVFSEKPR-UHFFFAOYSA-N
Synonym 2-n-pentylthiophene, thiophene, 2-pentyl, 2-n-amylthiophene, 1-methylbutylthiophene, unii-l79los1zkm, l79los1zkm, 2-amylthiophene, 2-pentyl thiophene, 2-pentyl-thiophene, acmc-209kdh
PubChem CID 20995
IUPAC Name 2-pentylthiophene
SMILES CCCCCC1=CC=CS1

Specifications

Boiling Point 204°C
Quantity 5 g
Formula Weight 154.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentylthiophene