missing translation for 'onlineSavingsMsg'
Learn More

2-Pentene (cis- and trans- mixture) 95.0+%, TCI America™

Supplier:  TCI America P0064100ML

 View more versions of this product

Catalog No. P0064100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Pentene (cis- and trans- mixture)
Colorless
C5H10
100 mL
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
CC\C=C/C
70.14
70.14
Liquid
109-68-2
36°C
MFCD00009384
3295
QMMOXUPEWRXHJS-HYXAFXHYSA-N
(2Z)-pent-2-ene
5326161
≥95.0% (GC)

Chemical Identifiers

109-68-2
70.14
QMMOXUPEWRXHJS-HYXAFXHYSA-N
5326161
CC\C=C/C
C5H10
MFCD00009384
trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
(2Z)-pent-2-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0019

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions