missing translation for 'onlineSavingsMsg'
Learn More

2-Pentene (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. P006425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P006425ML 25 mL
P0064100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P006425ML Supplier TCI America Supplier No. P006425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-68-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
IUPAC Name (2Z)-pent-2-ene
SMILES CC\C=C/C

Specifications

Color Colorless
Boiling Point 36°C
Quantity 25 mL
UN Number 3295
Formula Weight 70.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentene (cis- and trans- mixture)
EINECSNumber (2)-0019
TSCA Yes