missing translation for 'onlineSavingsMsg'
Learn More

2-Pentene, cis + trans, 98%

Catalog No. aaa1773614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1773614 25 g
AAA1773622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1773614 Supplier Thermo Scientific Chemicals Supplier No. A1773614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-68-2 | C5H10 | 70.14 g/mol

2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentene (mixture of cis and trans) is used in the synthesis of polycyclopentene by ring-opening metathesis polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.

Chemical Identifiers

CAS 109-68-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
SMILES CC\C=C/C

Specifications

Density 0.65
Boiling Point 36°C to 38°C
Flash Point −45°C (−49°F)
Odor Petroleum-like
Refractive Index 1.38
Linear Formula CH3CH2CH=CHCH3
Quantity 25 g
UN Number UN3295
Beilstein 969151
Merck Index 14,7123
Solubility Information Not miscible or difficult to mix in water.
IUPAC Name (2Z)-pent-2-ene
Formula Weight 70.14
Percent Purity 98%
Chemical Name or Material 2-Pentene, cis + trans
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 203-695-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only