Learn More
2-Pentanone, 99+%
CAS: 107-87-9 | C5H10O | 86.134 g/mol
Supplier: Thermo Scientific Chemicals L1326222
Description
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and reducing agents.
Specifications
| 2-Pentanone | |
| -78°C | |
| 100°C to 101°C | |
| C5H10O | |
| MFCD00009400 | |
| UN1249 | |
| 14,6114 | |
| Miscible with water. | |
| CCCC(=O)C | |
| 86.134 | |
| CHEBI:16472 | |
| ≥99% |
| 107-87-9 | |
| 0.813 | |
| 7°C (44°F) | |
| 1.3905 | |
| 100 g | |
| 506058 | |
| 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone | |
| XNLICIUVMPYHGG-UHFFFAOYSA-N | |
| pentan-2-one | |
| 7895 | |
| 86.13 |
Chemical Identifiers
| 107-87-9 | |
| 86.134 | |
| XNLICIUVMPYHGG-UHFFFAOYSA-N | |
| 7895 | |
| pentan-2-one |
| C5H10O | |
| MFCD00009400 | |
| 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone | |
| CHEBI:16472 | |
| CCCC(=O)C |
Safety and Handling
GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
H225-H302-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPYL KETONE
EINECSNumber : 203-528-1
RTECSNumber : SA7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only