missing translation for 'onlineSavingsMsg'
Learn More

2-Pentanone, 99+%

Catalog No. AAL1326214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1326214 25 g
AAL1326222 100 g
AAL1326236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1326214 Supplier Thermo Scientific Chemicals Supplier No. L1326214

CAS: 107-87-9 | C5H10O | 86.134 g/mol

2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pentanone is used as a solvent for lacquers and surface coatings. It acts as a flavoring agent. It is also used as a solvent for cleaning and degreasing purpose. Further, it serves as an intermediate, paint additive and coating additive.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 107-87-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00009400
InChI Key XNLICIUVMPYHGG-UHFFFAOYSA-N
Synonym 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone
PubChem CID 7895
ChEBI CHEBI:16472
IUPAC Name pentan-2-one
SMILES CCCC(=O)C

Specifications

Melting Point -78°C
Density 0.813
Boiling Point 100°C to 101°C
Flash Point 7°C (44°F)
Refractive Index 1.3905
Quantity 25 g
UN Number UN1249
Beilstein 506058
Merck Index 14,6114
Solubility Information Miscible with water.
Formula Weight 86.13
Percent Purity ≥99%
Chemical Name or Material 2-Pentanone
Show More Show Less
Hazard Category H225-H302-H319
Hazard Statement GHS H Statement
H225-H302-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPYL KETONE
EINECSNumber 203-528-1
RTECSNumber SA7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only