Learn More
2-Pentanone, 99+%, purified by redistillation, AcroSeal™
CAS: 107-87-9 | C5H10O | 86.13 g/mol
Supplier: Thermo Scientific Chemicals 224130010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Pentanone | |
| -78.0°C | |
| 100.0°C to 103.0°C | |
| 99% min. (GC) | |
| C5H10O | |
| CH3CO(CH2)CH3 | |
| 01,676 | |
| 15,6187 | |
| Solubility in water: 43g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| CCCC(=O)C | |
| 86.13 | |
| 0.5 mPa.s (20°C) | |
| 86.13 |
| 107-87-9 | |
| 0.8090g/mL | |
| 7°C | |
| AcroSeal™ Glass bottle | |
| 1.3890 to 1.3910 | |
| 1 L | |
| 0.809 | |
| 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone | |
| XNLICIUVMPYHGG-UHFFFAOYSA-N | |
| pentan-2-one | |
| 7895 | |
| CHEBI:16472 | |
| 99+% |
Chemical Identifiers
| 107-87-9 | |
| 86.13 | |
| 2-pentanone, methyl propyl ketone, ethyl acetone, pentanone, ethylacetone, propyl methyl ketone, metylopropyloketon, methyl n-propyl ketone, methylpropyl ketone, methyl-propyl-cetone | |
| CHEBI:16472 | |
| CCCC(=O)C |
| C5H10O | |
| XNLICIUVMPYHGG-UHFFFAOYSA-N | |
| 7895 | |
| pentan-2-one |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-528-1
RUO – Research Use Only