missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentanol 98.0+%, TCI America™
$59.65 - $262.83
Chemical Identifiers
| CAS | 6032-29-7 |
|---|---|
| Molecular Formula | C5H12O |
| Molecular Weight (g/mol) | 88.15 |
| MDL Number | MFCD00004579 |
| InChI Key | JYVLIDXNZAXMDK-UHFFFAOYSA-N |
| Synonym | 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol |
| PubChem CID | 22386 |
| ChEBI | CHEBI:77518 |
| IUPAC Name | pentan-2-ol |
| SMILES | CCCC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P005625ML
|
TCI America
P005625ML |
25 mL |
Each for $59.65
|
|
|||||
|
P0056500ML
|
TCI America
P0056500ML |
500 mL |
Each for $262.83
|
|
|||||
Chemical Identifiers
| 6032-29-7 | |
| 88.15 | |
| JYVLIDXNZAXMDK-UHFFFAOYSA-N | |
| 22386 | |
| pentan-2-ol |
| C5H12O | |
| MFCD00004579 | |
| 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol | |
| CHEBI:77518 | |
| CCCC(C)O |
Specifications
| 6032-29-7 | |
| 118°C | |
| MFCD00004579 | |
| 1105 | |
| JYVLIDXNZAXMDK-UHFFFAOYSA-N | |
| pentan-2-ol | |
| 22386 | |
| 88.15 | |
| Liquid |
| Colorless | |
| C5H12O | |
| 25 mL | |
| 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol | |
| CCCC(C)O | |
| 88.15 | |
| CHEBI:77518 | |
| ≥98.0% (GC) | |
| 2-Pentanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : SA4900000
TSCA : Yes