missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Pentanol, 99%

CAS: 6032-29-7 | C5H12O | 88.15 g/mol

$127.99 - $458.97

Chemical Identifiers

CAS 6032-29-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004579
InChI Key JYVLIDXNZAXMDK-UHFFFAOYSA-N
Synonym 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
PubChem CID 22386
ChEBI CHEBI:77518
IUPAC Name pentan-2-ol
SMILES CCCC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB21217AP
View Documents
Thermo Scientific Chemicals
B21217AP
500 mL
Each for $127.99
Only null left
 
AAB212170F
View Documents
Thermo Scientific Chemicals
B212170F
2500 mL
Each for $458.97
Only null left
 
Description

Description

It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

6032-29-7
88.15
JYVLIDXNZAXMDK-UHFFFAOYSA-N
22386
pentan-2-ol
C5H12O
MFCD00004579
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
CHEBI:77518
CCCC(C)O

Specifications

6032-29-7
0.811
33°C (91°F)
C5H12O
MFCD00004579
UN1105
14,7119
Soluble in water at 20°C,166g/L. Soluble in alcohol.
CCCC(C)O
88.15
CHEBI:77518
99%
(±)-2-Pentanol
°C to 50°C
114°C to 115°C
0.99
1.406
500 mL
1718819
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
JYVLIDXNZAXMDK-UHFFFAOYSA-N
pentan-2-ol
22386
88.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H335
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

EINECSNumber : 227-907-6

RTECSNumber : SA4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only