missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Pentanol, 99%

Catalog No. AAB212170F
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB212170F 2500 mL
AAB21217AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB212170F Supplier Thermo Scientific Chemicals Supplier No. B212170F
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6032-29-7 | C5H12O | 88.15 g/mol

It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.

Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6032-29-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004579
InChI Key JYVLIDXNZAXMDK-UHFFFAOYSA-N
Synonym 2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
PubChem CID 22386
ChEBI CHEBI:77518
IUPAC Name pentan-2-ol
SMILES CCCC(C)O

Specifications

Melting Point -50°C
Density 0.811
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Odor Characteristic
Assay Percent Range 0.99
Refractive Index 1.406
Quantity 2500 mL
UN Number UN1105
Beilstein 1718819
Merck Index 14,7119
Solubility Information Soluble in water at 20°C,166g/L. Soluble in alcohol.
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (+/-)-2-Pentanol
Show More Show Less
Hazard Category H226-H332-H335
Hazard Statement GHS H Statement
H226-H332-H335
Flammable liquid and vapour.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
EINECSNumber 227-907-6
RTECSNumber SA4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only