missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentadecanone, 97%
Description
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.
Solubility
Soluble in toluene (almost transparency). Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is primarily used as a flavoring substances to impart a fatty, slightly spicy, and floral flavor. 2-Pentadecanone can perfectly compliment and enhance a variety of foods including butters, cheeses and meats.
Solubility
Soluble in toluene (almost transparency). Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 37°C to 41°C |
| Density | 0.82 |
| Boiling Point | 293°C to 294°C |
| Flash Point | 144°C (291°F) |
| Quantity | 25 g |
| Beilstein | 1764872 |
| Solubility Information | Soluble in toluene (almost transparency). Insoluble in water. |
| Formula Weight | 226.4 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Pentadecanone |
Safety and Handling
| EINECSNumber | 219-064-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only