missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ 2-Part Female Pelvis Section Model

Model of Female Pelvis section including female genital organs with bladder and rectum fully exposed and removable.

Supplier:  Eisco™ AM0116B

Catalog No. S80798


Only null left
Explore available promotions

Description

Description

  • Female genital organs
  • Bladder and rectum fully exposed and removable
  • Separates into 2 parts
  • On Base
  • includes English Key Card
  • Dimensions: 35 x 33 x 10 cm
  • Weight: 1560 grams
Specifications

Specifications

2-part Model
Removable Female Reproductive Organs
1560 g
35 x 33 x 10 cm
PVC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions