missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)pyridine, 98%

Catalog No. AAH2657406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2657406 5 g
AAH2657414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2657406 Supplier Thermo Scientific Chemicals Supplier No. H2657406
Only null left

CAS: 4467-06-5 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4467-06-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00006281
InChI Key KJNZQKYSNAQLEO-UHFFFAOYSA-N
Synonym 2-p-tolyl pyridine, 2-4-methylphenyl pyridine, 2-p-tolylpyridine, 2-4-tolyl pyridine, pyridine, 2-4-methylphenyl, pyridine,2-p-tolyl, 2-p-toluyl pyridine, pubchem16062, 4-2-pyridyl toluene
PubChem CID 78219
IUPAC Name 2-(4-methylphenyl)pyridine
SMILES CC1=CC=C(C=C1)C2=CC=CC=N2

Specifications

Density 0.99
Boiling Point 170°C to 180°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 5 g
Formula Weight 169.22
Percent Purity 98%
Chemical Name or Material 2-(p-Tolyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-734-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only