missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)indole 98.0+%, TCI America™

Catalog No. T33511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33511G 1 g
T33515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33511G Supplier TCI America Supplier No. T33511G
Only null left

Chemical Identifiers

CAS 55577-25-8
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.28
MDL Number MFCD00087262
InChI Key VPXGIHGJJJBJFP-UHFFFAOYSA-N
Synonym 2-(4-Methylphenyl)indole
PubChem CID 292599
IUPAC Name 2-(4-methylphenyl)-1H-indole
SMILES CC1=CC=C(C=C1)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 1 g
Formula Weight 207.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Tolyl)indole