missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)ethanol 98.0+%, TCI America™

Catalog No. p-7133143
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22985G 5 g
M229825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22985G Supplier TCI America Supplier No. M22985G

Chemical Identifiers

CAS 699-02-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002904
InChI Key DAVFJRVIVZOKKS-UHFFFAOYSA-N
Synonym 2-4-methylphenyl ethanol, 2-p-tolyl ethanol, 4-methylphenethyl alcohol, benzeneethanol, 4-methyl, 2-p-tolylethanol, p-methylphenethyl alcohol, 4-methylbenzeneethanol, 2-4-methylphenyl ethan-1-ol, p-methylphenylethyl alcohol, 2-p-methylphenyl ethanol
PubChem CID 95294
IUPAC Name 2-(4-methylphenyl)ethanol
SMILES CC1=CC=C(C=C1)CCO

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(p-Tolyl)ethanol
EINECSNumber (3)-1068
TSCA Yes