missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(p-Tolyl)éthanol 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 115°C |
| Quantity | 5 g |
| Formula Weight | 136.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-(p-Tolyl)ethanol |
Sécurité et manipulation
| EINECSNumber | (3)-1068 |
| TSCA | Yes |