missing translation for 'onlineSavingsMsg'
Learn More

2-Oxovaleric Acid 95.0+%, TCI America™

Catalog No. p-7131743
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00151ML 1 mL
K00155ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00151ML Supplier TCI America Supplier No. K00151ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1821-02-9
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00066435
InChI Key KDVFRMMRZOCFLS-UHFFFAOYSA-N
Synonym 2-Ketopentanoic Acid, 2-Oxopentanoic Acid, 2-Ketovaleric Acid
PubChem CID 74563
ChEBI CHEBI:33033
IUPAC Name 2-oxopentanoic acid
SMILES CCCC(=O)C(O)=O

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 179°C
Quantity 1 mL
Formula Weight 116.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Oxovaleric Acid
EINECSNumber (2)-1503
TSCA No
Recommended Storage Refrigerator