missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxoglutaric Acid 99.0+%, TCI America™
$98.79 - $840.68
Chemical Identifiers
| CAS | 328-50-7 |
|---|---|
| Molecular Formula | C5H6O5 |
| Molecular Weight (g/mol) | 146.098 |
| MDL Number | MFCD00004165 |
| InChI Key | KPGXRSRHYNQIFN-UHFFFAOYSA-N |
| Synonym | 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid |
| PubChem CID | 51 |
| ChEBI | CHEBI:30915 |
| IUPAC Name | 2-oxopentanedioic acid |
| SMILES | C(CC(=O)O)C(=O)C(=O)O |
Chemical Identifiers
| 328-50-7 | |
| 146.098 | |
| KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
| 51 | |
| 2-oxopentanedioic acid |
| C5H6O5 | |
| MFCD00004165 | |
| 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
| CHEBI:30915 | |
| C(CC(=O)O)C(=O)C(=O)O |
Specifications
| 328-50-7 | |
| White-Yellow | |
| MFCD00004165 | |
| 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
| C(CC(=O)O)C(=O)C(=O)O | |
| 146.098 | |
| CHEBI:30915 | |
| ≥99.0% (T) | |
| 2-Oxoglutaric Acid |
| 116°C | |
| C5H6O5 | |
| 25 g | |
| KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
| 2-oxopentanedioic acid | |
| 51 | |
| 146.10 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0467
TSCA : Yes