missing translation for 'onlineSavingsMsg'
Learn More

(2-Oxo-3-benzoxazolyl)acetic acid, 97%

Catalog No. AAH6682806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6682806 5 g
AAH6682803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6682806 Supplier Thermo Scientific Chemicals Supplier No. H6682806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13610-49-6 | C9H7NO4 | 193.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13610-49-6
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD01547452
InChI Key PHIUXGVYFVAGTC-UHFFFAOYSA-N
Synonym 2-oxo-1,3-benzoxazol-3 2h-yl acetic acid, 2-oxo-benzooxazol-3-yl-acetic acid, 2-2-oxobenzo d oxazol-3 2h-yl acetic acid, 2 3h-benzoxazolone-3-yl acetic acid, 2-oxo-1,3-benzoxazol-3-yl acetic acid, 2-2-oxo-2,3-dihydro-1,3-benzoxazol-3-yl acetic acid, 2-2-oxo-3-hydrobenzoxazol-3-yl acetic acid, 3 2h-benzoxazoleacetic acid,2-oxo, 3-benzoxazolonyl acetic acid, tos-bb-0934
PubChem CID 737119
IUPAC Name 2-(2-oxo-1,3-benzoxazol-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)N(C(=O)O2)CC(=O)O

Specifications

Quantity 5 g
Beilstein 180971
Formula Weight 193.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (2-Oxo-3-benzoxazolyl)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only