missing translation for 'onlineSavingsMsg'
Learn More

2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC75703DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC75703DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

27685-90-1
233.62
IBHCNMJSVDRBSY-UHFFFAOYSA-N
119698
ClS(=O)(=O)C1=CC=C2NC(=O)OC2=C1
C7H4ClNO4S
MFCD03001119
2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 2-oxo-2,3-dihydrobenzo d oxazole-6-sulfonyl chloride, 6-benzoxazolesulfonyl chloride, 2,3-dihydro-2-oxo, 2-oxo-2,3-dihydro-benzooxazole-6-sulfonyl chloride, benzoxazalone, 6-sulfochloride, 2,3-dihydro-2-oxobenzoxazole-6-sulphonyl chloride, 2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonylchloride, 6-chlorosulfonyl-3-hydrobenzoxazol-2-one, pubchem5677, acmc-209gyq
2-oxo-3H-1,3-benzoxazole-6-sulfonyl chloride

Specifications

2-Oxo-2, 3-dihydro-1, 3-benzoxazole-6-sulfonyl chloride
94.5
95%
C7H4ClNO4S
MFCD03001119
IBHCNMJSVDRBSY-UHFFFAOYSA-N
2-oxo-3H-1,3-benzoxazole-6-sulfonyl chloride
119698
95%
27685-90-1
100.0
Amber Glass Bottle
5g
2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride, 2-oxo-2,3-dihydrobenzo d oxazole-6-sulfonyl chloride, 6-benzoxazolesulfonyl chloride, 2,3-dihydro-2-oxo, 2-oxo-2,3-dihydro-benzooxazole-6-sulfonyl chloride, benzoxazalone, 6-sulfochloride, 2,3-dihydro-2-oxobenzoxazole-6-sulphonyl chloride, 2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonylchloride, 6-chlorosulfonyl-3-hydrobenzoxazol-2-one, pubchem5677, acmc-209gyq
ClS(=O)(=O)C1=CC=C2NC(=O)OC2=C1
233.62
233.63
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents