missing translation for 'onlineSavingsMsg'
Learn More

2-Oxetanemethanol, 96%

Catalog No. p-7056965
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66218MD 250 mg
AAH6621803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66218MD Supplier Thermo Scientific Chemicals Supplier No. H66218MD
Only null left

CAS: 61266-70-4 | C4H8O2 | 88.106 g/mol

2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.

Chemical Identifiers

CAS 61266-70-4
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00142933
InChI Key PQZJTHGEFIQMCO-UHFFFAOYSA-N
Synonym 2-hydroxymethyloxetane, 2-oxetanemethanol, 2-oxetanylmethanol, 2,4-epoxybutanol, oxetan-2-yl-methanol, oxetane-2-methanol, methyloloxacyclobutan, 2-hydrpxymethyloxetane, oxetan-2-yl methanol, rac-2-hydroxymethyloxetane
PubChem CID 12284142
IUPAC Name oxetan-2-ylmethanol
SMILES C1COC1CO

Specifications

Color Colorless to Yellow
Density 1.080 g/mL
Boiling Point 98°C (15 mmHg)
Refractive Index 1.446
Quantity 250 mg
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 88.11
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Oxetanemethanol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only